Order Entry
Italy
ContactUsLinkComponent
 
1-Bromo-2-(2-methoxyethoxy)ethane ≥90.0% (by GC) stabilised
 :  TCI
 
1-Bromo-2-(2-methoxyethoxy)ethane ≥90.0% (by GC) stabilised
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 90.0 %

 

Frequently Bought Together