
3-(tert-Butoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:220210-56-0
:
Formula:C₁₁H₁₅BO₄
MDL Number:MFCD01630855
:222.05 g/mol
Melting Pt:102°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 109.0 %
Specification Test Results
Related Information
Frequently Bought Together