Order Entry
Italy
ContactUsLinkComponent
4,7-Bis(5-bromo-4-dodecyl-2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC)
4,7-Bis(5-bromo-4-dodecyl-2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC)
Catalog # TCIAB4719-200MG
Supplier:  TCI
CAS Number:  
4,7-Bis(5-bromo-4-dodecyl-2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC)
Catalog # TCIAB4719-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Orange to Red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 95.0 %

Specification Test Results

About this item

 :
 :794.85 g/mol
CAS Number:1179993-72-6
Formula:C₃₈H₅₄Br₂N₂S₃
MDL Number:MFCD27923036