

4,8-Bis(2-butyl-n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)
Catalog # TCIAB4685-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Sulfuride):6.70 to 7.60 %
Specification Test Results
About this item
:
:884.54 g/mol
UN:3146
ADR:6.1,I
:GHS06
:GHS09
CAS Number:1271439-08-7
Formula:C₄₀H₇₀O₂S₂Sn₂