4,7-Bis(5-n-octyl-2-thienyl)-2,1,3-benzothiadiazole ≥98.0% (by HPLC)
Catalog # TCIAB4683-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):5.00 to 5.60 %
Specification Test Results
About this item
:
:524.84 g/mol
CAS Number:1171974-28-9
Formula:C₃₀H₄₀N₂S₃
MDL Number:MFCD28976158