
6-Bromo-m-toluonitrile ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:42872-83-3
:
Formula:C₈H₆BrN
MDL Number:MFCD11040281
:196.05 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
Frequently Bought Together