
3-(4-Bromophenyl)propan-1-ol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25574-11-2
:
Formula:C₉H₁₁BrO
MDL Number:MFCD09028724
:215.09 g/mol
Boiling Pt:162°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together