Order Entry
Italy
ContactUsLinkComponent
 
 
3-(4-Bromophenyl)pyridine ≥96.0% (by GC)
 :  TCI
 
 
3-(4-Bromophenyl)pyridine ≥96.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    38.0 to 42.0 deg-C

 

Frequently Bought Together