N,N'-Bis[2-(dimethylamino)ethyl]-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB4583-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:231.0 to 235.0 deg-C
Specification Test Results
About this item
:
:408.46 g/mol
CAS Number:22291-04-9
Formula:C₂₂H₂₄N₄O₄
MDL Number:MFCD27923061