Order Entry
Italy
ContactUsLinkComponent
 
9-Bromo-10-(9-phenanthryl)anthracene ≥98.0% (by HPLC)
 :  TCI
 
9-Bromo-10-(9-phenanthryl)anthracene ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 95.0 %
  • Melting point:
    231.0 to 236.0 deg-C

 

Frequently Bought Together