Order Entry
Italy
ContactUsLinkComponent
2,2'-Bipyridine-6-carbonitrile ≥98.0% (by GC)
Irritant
2,2'-Bipyridine-6-carbonitrile ≥98.0% (by GC)
Catalog # TCIAB4557-200MG
Supplier:  TCI
CAS Number:  
Irritant
2,2'-Bipyridine-6-carbonitrile ≥98.0% (by GC)
Catalog # TCIAB4557-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    131.0 to 135.0 deg-C

Specification Test Results

About this item

 :
 :181.20 g/mol
 :GHS07
CAS Number:4392-85-2
Formula:C₁₁H₇N₃
MDL Number:MFCD07364147