
2H-1,4-Benzoxazin-3(4H)-one ≥98.0% (by GC)
: TCI
:5466-88-6
:
:C₈H₇NO₂
:MFCD00158536
:149.15 g/mol
:GHS07
- Appearance:White to Orange to Green clear liquid
- Purity (GC):min. 98.0 %
- Melting point:172.0 to 176.0 deg-C
Frequently Bought Together