4-[Bis(4-iodophenyl)amino]benzaldehyde ≥95.0% (by GC)
TCIAB4543-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:139.0 to 143.0 deg-C
:
:525.13 g/mol
:808758-81-8
:C₁₉H₁₃I₂NO
:MFCD20259247