Order Entry
Italy
ContactUsLinkComponent
 
 
2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
 :  TCI
 
 
2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together