Order Entry
Italy
ContactUsLinkComponent
5,6-Bis(n-octyloxy)-2,1,3-benzothiadiazole ≥98.0% (by GC)
5,6-Bis(n-octyloxy)-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAB4440-200MG
Supplier:  TCI
CAS Number:  
5,6-Bis(n-octyloxy)-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAB4440-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    94.0 to 98.0 deg-C

Specification Test Results

About this item

 :
 :392.60 g/mol
CAS Number:1254353-37-1
Formula:C₂₂H₃₆N₂O₂S
MDL Number:MFCD19440987