Order Entry
Italy
ContactUsLinkComponent
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) sodium salt ≥85.0% (by HPLC)
Irritant
Health
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) sodium salt ≥85.0% (by HPLC)
Catalog # TCIAB4432-200MG
Supplier:  TCI
CAS Number:  
Irritant
Health
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) sodium salt ≥85.0% (by HPLC)
Catalog # TCIAB4432-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Purity (HPLC):
    min. 85.0 area%
  • Elemental analysis (Nitrogen):
    8.40 to 9.70 %

Specification Test Results

About this item

 :
 :892.09 g/mol
 :GHS07
 :GHS08
CAS Number:871466-65-8
Formula:C₄₂H₅₁N₆NaO₄RuS₂