Order Entry
Italy
ContactUsLinkComponent
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)
Catalog # TCIAB4422-50MG
Supplier:  TCI
CAS Number:  
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)
Catalog # TCIAB4422-50MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

About this item

 :
 :1073.84 g/mol
Melting Pt:285°C
CAS Number:1253900-41-2
Formula:C₇₆H₁₀₄Si₂