Order Entry
Italy
ContactUsLinkComponent
(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥70% (by HPLC)
(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥70% (by HPLC)
Catalog # TCIAB4421-10MG
Supplier:  TCI
CAS Number:  
(E)-1,2-Bis(1-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥70% (by HPLC)
Catalog # TCIAB4421-10MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 70.0 area%

Specification Test Results

About this item

 :
 :1073.84 g/mol
CAS Number:1620487-87-7
Formula:C₇₆H₁₀₄Si₂