Order Entry
Italy
ContactUsLinkComponent
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis)
Irritant
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Irritant
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    88.0 to 92.0 deg-C

Specification Test Results

Frequently Bought Together