Order Entry
Italy
ContactUsLinkComponent
4,8-Bis(n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
Toxic
Environment
4,8-Bis(n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
Catalog # TCIAB4378-200MG
Supplier:  TCI
CAS Number:  
Toxic
Environment
4,8-Bis(n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene ≥95.0% (by HPLC)
Catalog # TCIAB4378-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Elemental analysis (Sulfuride):
    7.70 to 8.70 %

Specification Test Results

About this item

 :
 :772.32 g/mol
UN:3146
ADR:6.1,I
 :GHS06
 :GHS09
CAS Number:1098102-95-4
Formula:C₃₂H₅₄O₂S₂Sn₂