Order Entry
Italy
ContactUsLinkComponent
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) ≥80.0% (by HPLC)
Health
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) ≥80.0% (by HPLC)
Catalog # TCIAB4373-200MG
Supplier:  TCI
CAS Number:  
Health
Bis(isothiocyanato)(2,2'-bipyridyl-4,4'-dicarboxylato)(4,4'-dinonyl-2,2'-bipyridyl)ruthenium(II) ≥80.0% (by HPLC)
Catalog # TCIAB4373-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Purity (HPLC):
    min. 80.0 area%
  • Purity (Neutralization titration):
    min. 80.0 %
  • Elemental analysis (Nitrogen):
    8.00 to 10.00 %

Specification Test Results

About this item

 :
 :870.11 g/mol
 :GHS08
CAS Number:502693-09-6
Formula:C₄₂H₅₂N₆O₄RuS₂
MDL Number:MFCD12546029