Order Entry
Italy
ContactUsLinkComponent
2-Bromo-9,9-di-n-octylfluorene ≥97.0% (by GC)
Irritant
2-Bromo-9,9-di-n-octylfluorene ≥97.0% (by GC)
Catalog # TCIAB4366-1G
Supplier:  TCI
CAS Number:  
Irritant
2-Bromo-9,9-di-n-octylfluorene ≥97.0% (by GC)
Catalog # TCIAB4366-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :469.55 g/mol
 :GHS07
CAS Number:302554-80-9
Formula:C₂₉H₄₁Br
MDL Number:MFCD12024273