Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-4-((methylsulphonyl)oxy)piperidine-1-carboxylate ≥98.0% (by GC)
Irritant
tert-Butyl-4-((methylsulphonyl)oxy)piperidine-1-carboxylate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
tert-Butyl-4-((methylsulphonyl)oxy)piperidine-1-carboxylate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together