
3,3'-Bipiperidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4309-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
Melting Pt:105°C
:168.28 g/mol
:GHS07
CAS Number:46040-83-9
Formula:C₁₀H₂₀N₂