
N,N',N''-Trimethyldiethylenetriamine ≥95.0% (by GC, titration analysis)
TCIAB4304-5ML
: TCI
:
- Pk:5 mL
- Seq:2210
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
:
:145.25 g/mol
:56°C
:203-337-3
:2735
:8,II
:GHS05
:105-84-0
:C₇H₁₉N₃
:MFCD00025637