
N-Boc-N-methylethylenediamine ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:121492-06-6
:
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD01317789
:174.24 g/mol
Boiling Pt:79°C
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together