Order Entry
Italy
ContactUsLinkComponent
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    190.0 to 194.0 deg-C

Specification Test Results

Frequently Bought Together