
2,4-Bis(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
: TCI
:59664-42-5
:
:C₉H₄F₆O
:MFCD00083535
:242.12 g/mol
:68°C
:GHS07
:Refrigerator
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together