
Biphenyl-3-carboxaldehyde ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1204-60-0
:
Formula:C₁₃H₁₀O
MDL Number:MFCD01740432
:182.22 g/mol
Boiling Pt:125°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together