
2-Butyl-3-[[2'-cyano-[1,1'-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4,4]non-1-en-4-one ≥97.0% (by total nitrogen basis)
: TCI
:138401-24-8
:
:C₂₅H₂₇N₃O
:MFCD06658242
:385.51 g/mol
:423-500-4
:3077
:9,III
:GHS09
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:91.0 to 95.0 deg-C
Frequently Bought Together