
2-Bromo-4,5-diluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:875664-38-3
:
Formula:C₇H₅BrF₂
MDL Number:MFCD07777160
:207.02 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together