
2,2'-Dibromoacetophenone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:49851-55-0
:
Formula:C₈H₆Br₂O
MDL Number:MFCD00044660
:277.9406 g/mol
Boiling Pt:100°C
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together