Order Entry
Italy
ContactUsLinkComponent
1,2-Bis(triethoxysilyl)ethane ≥95.0% (by GC)
Corrosive
1,2-Bis(triethoxysilyl)ethane ≥95.0% (by GC)
Catalog # TCIAB4123-5ML
Supplier:  TCI
CAS Number:  
Corrosive
1,2-Bis(triethoxysilyl)ethane ≥95.0% (by GC)
Catalog # TCIAB4123-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :354.59 g/mol
Boiling Pt:88°C
EINECS:240-212-2
 :GHS05
CAS Number:16068-37-4
Formula:C₁₄H₃₄O₆Si₂
MDL Number:MFCD00239362