4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:365547-20-2
:
Formula:C₂₅H₃₈S₂
MDL Number:MFCD28291964
:402.70 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together