Order Entry
Italy
ContactUsLinkComponent
1,2-Butanedithiol ≥97.0% (by GC)
Irritant
1,2-Butanedithiol ≥97.0% (by GC)
Catalog # TCIAB4071-5G
Supplier:  TCI
CAS Number:  
Irritant
1,2-Butanedithiol ≥97.0% (by GC)
Catalog # TCIAB4071-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :122.26 g/mol
Boiling Pt:77°C
EINECS:240-290-8
 :GHS07
CAS Number:16128-68-0
Formula:C₄H₁₀S₂
MDL Number:MFCD00039649