
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19184-65-7
:
Formula:C₅H₁₁BrO₃
MDL Number:MFCD00135306
:199.04 g/mol
EINECS:242-863-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:75.0 to 79.0 deg-C
Specification Test Results
Frequently Bought Together