Order Entry
Italy
ContactUsLinkComponent
Ethyl palmitate for synthesis, Sigma-Aldrich®
Ethyl palmitate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ethyl palmitate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    23 - 26 °C
  • Molar Mass:
    284.48 g/mol
  • Boiling Point:
    192 - 194 °C (13 hPa)
  • Flash Point:
    164 °C
  • Refractive Index:
    1.438 (25 °C, 589 nm)
  • Density:
    0.86 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together