
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:667463-62-9
:
Formula:C₁₆H₁₀BrN₃O₂
MDL Number:MFCD08705318
Melting Pt:300°C
:356.18 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):11.30 to 12.30 %
Specification Test Results
Frequently Bought Together