Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥97.0% (by HPLC) for biochemical research
Catalog # TCIAB4000-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):7.80 to 8.40 %
- Specific rotation [a]20/D:-12.0 to -14.0 deg (C=0.5、HCl)
- Functionality test:to pass test
Specification Test Results
About this item
:
:308.4 · 36.5 g/mol
Storage Temperature:Freezer
CAS Number:65391-42-6
Formula:C₁₆H₂₅ClN₂O₄
MDL Number:MFCD00058004