3,3'-Bipyridyl ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:581-46-4
:
Formula:C₁₀H₈N₂
MDL Number:MFCD00006371
:156.19 g/mol
Boiling Pt:175°C
EINECS:209-466-1
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together