Order Entry
Italy
ContactUsLinkComponent
 
 
(11bS)-2,6-Bis[bis[3,5-bis(trifluoromethyl)phenyl]hydroxymethyl]-3,5-dihydrospiro[4H-dinaphth[2,1-c:1',2'-e]azepine-4,4'-morpholinium] bromide ≥97.0% (by HPLC)
  TCIAB3970-50MG
 :  TCI
 :  
 
(11bS)-2,6-Bis[bis[3,5-bis(trifluoromethyl)phenyl]hydroxymethyl]-3,5-dihydrospiro[4H-dinaphth[2,1-c:1',2'-e]azepine-4,4'-morpholinium] bromide ≥97.0% (by HPLC)
  TCIAB3970-50MG
 :  TCI
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

 

 

 :
 :1354.82 g/mol
 :GHS07
 :1197922-04-5
 :C₆₀H₃₆BrF₂₄NO₃