
(2-Bromoethyl)diphenylsulphonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:247129-85-7
:
Formula:C₁₅H₁₄BrF₃O₃S₂
MDL Number:MFCD12546049
:443.29 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
Specification Test Results
Frequently Bought Together