Order Entry
Italy
ContactUsLinkComponent
1,1'-Binaphthyl-2,2'-diamine ≥98.0% (by HPLC, titration analysis)
1,1'-Binaphthyl-2,2'-diamine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3923-1G
Supplier:  TCI
CAS Number:  
1,1'-Binaphthyl-2,2'-diamine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3923-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    190.0 to 194.0 deg-C

Specification Test Results

About this item

 :
 :284.36 g/mol
CAS Number:4488-22-6
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00145204