Order Entry
Italy
ContactUsLinkComponent
1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol ≥98.0% (by GC)
1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol ≥98.0% (by GC)
  TCIAB3905-100MG
 :  TCI
 :  
1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol ≥98.0% (by GC)
  TCIAB3905-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    50.0 to 55.0 deg-C

 

 

 :
 :320.38 g/mol
 :Refrigerator
 :847682-00-2
 :C₁₅H₂₈O₇