2-(5-Bromo-2-thienyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine ≥98.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:927384-46-1
:
Formula:C₁₄H₁₀BBrN₂S
:329.02 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together