Order Entry
Italy
ContactUsLinkComponent
Biphenyl-3,3',5,5'-tetracarboxylic acid ≥98.0% (by HPLC)
Irritant
Biphenyl-3,3',5,5'-tetracarboxylic acid ≥98.0% (by HPLC)
Catalog # TCIAB3792-200MG
Supplier:  TCI
CAS Number:  
Irritant
Biphenyl-3,3',5,5'-tetracarboxylic acid ≥98.0% (by HPLC)
Catalog # TCIAB3792-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :330.25 g/mol
 :GHS07
CAS Number:4371-28-2
Formula:C₁₆H₁₀O₈
MDL Number:MFCD16621475