Order Entry
Italy
ContactUsLinkComponent
Bis[2-(4-azidosalicylamido)ethyl]disulphide ≥97.0% (by HPLC)
Bis[2-(4-azidosalicylamido)ethyl]disulphide ≥97.0% (by HPLC)
Catalog # TCIAB3790-10MG
Supplier:  TCI
CAS Number:  
Bis[2-(4-azidosalicylamido)ethyl]disulphide ≥97.0% (by HPLC)
Catalog # TCIAB3790-10MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Elemental analysis (Nitrogen):
    22.90 to 23.90 %

Specification Test Results

About this item

 :
 :474.51 g/mol
Storage Temperature:Refrigerator
CAS Number:199804-21-2
Formula:C₁₈H₁₈N₈O₄S₂
MDL Number:MFCD01861964