Order Entry
Italy
ContactUsLinkComponent
5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Irritante
5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Irritante
5-Bromo-2-oxo-1,2-dihydropyridine-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Risultati dei test sulle specifiche

Frequently Bought Together