Order Entry
Italy
ContactUsLinkComponent
(2S,3S,5R,6R)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥98.0% (by GC)
(2S,3S,5R,6R)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥98.0% (by GC)
Catalog # TCIAB3696-200MG
Supplier:  TCI
CAS Number:  
(2S,3S,5R,6R)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥98.0% (by GC)
Catalog # TCIAB3696-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    118.0 to 122.0 deg-C
  • Specific rotation [a]20/D:
    +172 to +176 deg (C=1, CHCl3)

Specification Test Results

About this item

 :
 :236.26 g/mol
CAS Number:241811-66-5
Formula:C₁₀H₂₀O₆