(2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥98.0% (by GC)
Catalog # TCIAB3695-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:119.0 to 123.0 deg-C
- Specific rotation [a]20/D:-172 to -176 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:236.26 g/mol
CAS Number:173371-55-6
Formula:C₁₀H₂₀O₆